Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 10 - 11 | 1511 | 67 | |
GenBank mRNA | AF093576.1 | - | 10 - 11 | 1419 | 67 | |
AK142860.1 | - | 10 - 11 | 1486 | 67 | ||
BC150747.1 | - | 10 - 11 | 1376 | 9087658 | 67 | |
U87455.1 | - | 10 - 11 | 1491 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |