Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007424.2 | NP_031450.2 | 10 - 11 | 2186 | 57 | |
GenBank mRNA | L07049.1 | - | 10 - 11 | 2185 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |