Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026231.2 | NP_080507.2 | 7 - 8 | 1585 | 89 | |
GenBank mRNA | AK013967.1 | - | 4 - 5 | 437 | 89 | |
AK017059.1 | - | 5 - 6 | 860 | 89 | ||
AK020021.1 | - | 6 - 7 | 1195 | 89 | ||
AK029724.1 | - | 7 - 8 | 1585 | 89 | ||
AK033409.1 | - | 7 - 8 | 1623 | 89 | ||
AK044441.1 | - | 7 - 8 | 1593 | 89 | ||
BC047136.1 | - | 7 - 8 | 1585 | 6311866 | 89 | |
BC061240.1 | - | 7 - 8 | 1581 | 6743615 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |