Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083312.2 | NP_001076781.1 | 2 - 3 | 307 | 69 | |
NM_145545.4 | NP_663520.2 | 2 - 3 | 325 | 69 | ||
GenBank mRNA | AK036666.1 | - | 2 - 3 | 321 | 69 | |
AK087751.1 | - | 2 - 3 | 301 | 69 | ||
AK131176.1 | - | 1 - 2 | 208 | 69 | ||
BC010229.1 | - | 2 - 3 | 272 | 3501441 | 69 | |
BY742706.1 | - | 2 - 3 | 305 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |