Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291184.1 | NP_001278113.1 | 1 - 2 | 301 | 80 | |
NM_011048.1 | NP_035178.1 | 1 - 2 | 301 | 80 | ||
GenBank mRNA | D50060.1 | - | 1 - 2 | 224 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |