Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tatdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163421.1 | NP_001156893.1 | 5 - 6 | 391 | 128 | |
NM_001163422.1 | NP_001156894.1 | 5 - 6 | 391 | 128 | ||
NM_026895.1 | NP_081171.1 | 5 - 6 | 391 | 128 | ||
GenBank mRNA | AK005193.1 | - | 5 - 6 | 362 | 128 | |
AK156071.1 | - | 5 - 6 | 342 | 128 | ||
BC061248.1 | - | 5 - 6 | 363 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |