Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172092.1 | NP_001165563.1 | 1 - 2 | 139 | 74 | |
NM_001172093.1 | NP_001165564.1 | 1 - 2 | 139 | 74 | ||
NM_029523.3 | NP_083799.2 | 1 - 2 | 139 | 74 | ||
GenBank mRNA | AK050456.1 | - | 1 - 2 | 76 | 74 | |
AK088719.1 | - | 1 - 2 | 115 | 74 | ||
AK146458.1 | - | 1 - 2 | 110 | 74 | ||
BB857645.1 | - | 1 - 2 | 139 | 74 | ||
BC023934.1 | - | 1 - 2 | 132 | 5342167 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |