Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700030368.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030368.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030368.mm.1 |
TC0700003591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030366.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030366.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030366.mm.1 |
TC0700003591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030367.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030367.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030367.mm.1 |
TC0700003591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030369.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030369.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030369.mm.1 |
TC0700003591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030370.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030370.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030370.mm.1 |
TC0700003591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700030365.mm.1 |
NM_133952.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030365.mm.1 |
NM_133952.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700030365.mm.1 |
TC0700003591.mm.1 |
MTA TC |