Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100031133.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031133.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031133.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031126.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031126.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031126.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031128.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031128.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031128.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031134.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031134.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031134.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031129.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031129.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031129.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031131.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031131.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031131.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031127.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031127.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031127.mm.1 |
TC0100003806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100031130.mm.1 |
NM_144880.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031130.mm.1 |
NM_144880.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100031130.mm.1 |
TC0100003806.mm.1 |
MTA TC |