Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291222.1 | NP_001278151.1 | 10 - 11 | 2437 | 56 | |
NM_010626.3 | NP_034756.2 | 10 - 11 | 2437 | 56 | ||
GenBank mRNA | BC141045.1 | - | 10 - 11 | 2438 | 9055657 | 56 |
BC145222.1 | - | 10 - 11 | 2438 | 9053773 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |