Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027790.2 | NP_082066.2 | 4 - 5 | 369 | 59 | |
GenBank mRNA | AK017274.1 | - | 4 - 5 | 371 | 59 | |
BC117731.2 | - | 3 - 4 | 262 | 40089799 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |