Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027193.2 | NP_081469.2 | 6 - 7 | 652 | 76 | |
GenBank mRNA | AK010475.1 | - | 6 - 7 | 642 | 76 | |
BC060372.1 | - | 6 - 7 | 652 | 6391546 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |