Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017462.2 | NP_059490.2 | 17 - 18 | 3034 | 65 | |
GenBank mRNA | AB121698.1 | - | 16 - 17 | 2727 | 65 | |
AK133584.1 | - | 17 - 18 | 2960 | 65 | ||
AK148091.1 | - | 17 - 18 | 2996 | 65 | ||
AK162462.1 | - | 3 - 4 | 525 | 65 | ||
BC053024.1 | - | 17 - 18 | 2960 | 65 | ||
BC061472.1 | - | 17 - 18 | 2948 | 6827238 | 65 | |
BC080291.1 | - | 17 - 18 | 2865 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |