Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cysltr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133720.3 | NP_598481.2 | 3 - 4 | 244 | 134 | |
GenBank mRNA | AF331658.1 | - | 3 - 4 | 203 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus Probe spans exons |