Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146246.3 | NP_666358.2 | 2 - 3 | 857 | 68 | |
GenBank mRNA | AJ491325.1 | - | 1 - 2 | 659 | 68 | |
AY168340.1 | - | 1 - 2 | 638 | 68 | ||
BC129887.1 | - | 2 - 3 | 782 | 40111348 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |