Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172599.4 | NP_766187.1 | 3 - 4 | 361 | 69 | |
GenBank mRNA | AK029330.1 | - | 3 - 4 | 342 | 69 | |
AK029967.1 | - | 3 - 4 | 342 | 69 | ||
AK054196.1 | - | 3 - 4 | 362 | 69 | ||
BC057360.1 | - | 2 - 3 | 81 | 69 | ||
BC090995.1 | - | 3 - 4 | 341 | 6754513 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |