Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134076.2 | NP_598837.2 | 1 - 2 | 85 | 106 | |
GenBank mRNA | AK049366.1 | - | 1 - 2 | 66 | 106 | |
AK152595.1 | - | 1 - 2 | 86 | 106 | ||
AK161853.1 | - | 1 - 2 | 35 | 106 | ||
AK164655.1 | - | 1 - 2 | 45 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |