Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007576.2 | NP_001007577.1 | 6 - 7 | 1818 | 108 | |
GenBank mRNA | AK044234.1 | - | 6 - 7 | 1819 | 108 | |
AY651761.1 | - | 6 - 7 | 1813 | 108 | ||
BC115714.1 | - | 5 - 6 | 1648 | 40085764 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |