Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sardh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138665.2 | NP_619606.1 | 8 - 9 | 1822 | 77 | |
GenBank mRNA | AK140451.1 | - | 7 - 8 | 1098 | 77 | |
AK159236.1 | - | 8 - 9 | 1786 | 77 | ||
AK159681.1 | - | 8 - 9 | 1786 | 77 | ||
AK159704.1 | - | 8 - 9 | 1786 | 77 | ||
AK163515.1 | - | 8 - 9 | 1351 | 77 | ||
AK163660.1 | - | 7 - 8 | 1098 | 77 | ||
BC003456.1 | - | 8 - 9 | 1331 | 2648344 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |