Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027352.4 | NP_081628.3 | 6 - 7 | 891 | 130 | |
NR_027343.1 | 7 - 8 | 1088 | 130 | |||
GenBank mRNA | AK002515.1 | - | 7 - 8 | 969 | 129 | |
AK010664.2 | - | 6 - 7 | 810 | 130 | ||
AK033413.1 | - | 6 - 7 | 825 | 130 | ||
AK044691.1 | - | 776 | 130 | |||
AK077153.1 | - | 6 - 7 | 766 | 130 | ||
AK077683.1 | - | 6 - 7 | 826 | 130 | ||
AK077706.1 | - | 6 - 7 | 758 | 130 | ||
AK088239.1 | - | 6 - 7 | 772 | 130 | ||
AK148337.1 | - | 6 - 7 | 825 | 130 | ||
AK151843.1 | - | 6 - 7 | 773 | 130 | ||
AK152475.1 | - | 6 - 7 | 773 | 130 | ||
AK152544.1 | - | 6 - 7 | 773 | 130 | ||
AK152603.1 | - | 6 - 7 | 801 | 130 | ||
AK153049.1 | - | 6 - 7 | 773 | 130 | ||
AK153555.1 | - | 6 - 7 | 824 | 130 | ||
AK159469.1 | - | 6 - 7 | 823 | 130 | ||
AK167823.1 | - | 6 - 7 | 825 | 130 | ||
AK168928.1 | - | 7 - 8 | 1021 | 130 | ||
AK169382.1 | - | 6 - 7 | 773 | 130 | ||
AK184024.1 | - | 1 - 2 | 148 | 130 | ||
AK188100.1 | - | 1 - 2 | 149 | 131 | ||
AK197786.1 | - | 1 - 2 | 148 | 130 | ||
AK198884.1 | - | 1 - 2 | 148 | 130 | ||
AK216502.1 | - | 1 - 2 | 107 | 130 | ||
AK216984.1 | - | 1 - 2 | 148 | 130 | ||
AK218436.1 | - | 1 - 2 | 148 | 130 | ||
BC005600.1 | - | 6 - 7 | 764 | 3482776 | 130 | |
BC016455.1 | - | 5 - 6 | 616 | 4234343 | 130 | |
BC024982.1 | - | 6 - 7 | 777 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |