Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133207.2 | NP_573470.2 | 14 - 15 | 3342 | 99 | |
GenBank mRNA | AJ291608.1 | - | 14 - 15 | 3149 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons |