Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291039.1 | NP_001277968.1 | 9 - 10 | 799 | 96 | |
NM_138651.7 | NP_619592.1 | 9 - 10 | 1122 | 96 | ||
GenBank mRNA | AK036328.1 | - | 9 - 10 | 973 | 96 | |
AK147541.1 | - | 9 - 10 | 1014 | 96 | ||
AK166001.1 | - | 9 - 10 | 973 | 96 | ||
AY159802.1 | - | 9 - 10 | 1045 | 96 | ||
BC003852.1 | - | 9 - 10 | 1028 | 3600825 | 96 | |
BC069879.1 | - | 9 - 10 | 1001 | 30439069 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |