Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019456.2 | NP_062329.2 | 7 - 8 | 1017 | 136 | |
GenBank mRNA | AF020313.1 | - | 4 - 5 | 623 | 136 | |
AK021331.1 | - | 8 - 9 | 896 | 136 | ||
AK040084.1 | - | 7 - 8 | 953 | 136 | ||
AK042467.1 | - | 7 - 8 | 955 | 136 | ||
BC003991.1 | - | 4 - 5 | 668 | 136 | ||
BC023110.1 | - | 7 - 8 | 906 | 3707478 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |