Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007027.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007027.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007027.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007030.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007030.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007030.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007032.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007032.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007032.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007026.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007026.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007026.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007029.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007029.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007029.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007028.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007028.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007028.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007031.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007031.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007031.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007033.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007033.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007033.mm.1 |
TC1100000856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007034.mm.1 |
NM_177369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007034.mm.1 |
NM_177369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007034.mm.1 |
TC1100000856.mm.1 |
MTA TC |