Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029934.3 | NP_084210.2 | 4 - 5 | 1120 | 64 | |
GenBank mRNA | AB055410.1 | - | 4 - 5 | 1118 | 64 | |
AK019981.1 | - | 4 - 5 | 531 | 64 | ||
AK038998.1 | - | 4 - 5 | 542 | 64 | ||
AK044149.1 | - | 4 - 5 | 547 | 64 | ||
AK150010.1 | - | 4 - 5 | 514 | 64 | ||
AK154606.1 | - | 4 - 5 | 1119 | 64 | ||
AK158998.1 | - | 3 - 4 | 875 | 64 | ||
AK170124.1 | - | 4 - 5 | 597 | 64 | ||
BC023417.1 | - | 4 - 5 | 568 | 5028759 | 64 | |
BC118958.2 | - | 4 - 5 | 765 | 40093821 | 64 | |
EU016380.1 | - | 3 - 4 | 332 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |