Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013908.4 | NP_038936.1 | 7 - 8 | 1387 | 59 | |
GenBank mRNA | AF176520.1 | - | 7 - 8 | 1348 | 59 | |
AK019715.2 | - | 7 - 8 | 1388 | 59 | ||
AK154911.1 | - | 6 - 7 | 2326 | 59 | ||
AK188841.1 | - | 1 - 2 | 91 | 59 | ||
BC010776.1 | - | 7 - 8 | 1314 | 4211592 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |