Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028193.3 | NP_082469.2 | 5 - 6 | 810 | 67 | |
GenBank mRNA | BC037224.1 | - | 5 - 6 | 593 | 3257364 | 67 |
CF737042.1 | - | 2 - 3 | 131 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |