Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027014.1 | NP_081290.1 | 5 - 6 | 569 | 110 | |
GenBank mRNA | AK013116.1 | - | 5 - 6 | 562 | 110 | |
AK129081.1 | - | 4 - 5 | 360 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |