Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290563.1 | NP_001277492.1 | 5 - 6 | 749 | 63 | |
NM_001290564.1 | NP_001277493.1 | 5 - 6 | 804 | 63 | ||
NM_001290566.1 | NP_001277495.1 | 4 - 5 | 502 | 63 | ||
NM_001290567.1 | NP_001277496.1 | 5 - 6 | 550 | 63 | ||
NM_177384.3 | NP_796358.2 | 5 - 6 | 749 | 63 | ||
GenBank mRNA | AK028040.1 | - | 5 - 6 | 804 | 63 | |
AK089384.1 | - | 4 - 5 | 782 | 63 | ||
AK133273.1 | - | 5 - 6 | 527 | 63 | ||
AK160435.1 | - | 5 - 6 | 500 | 63 | ||
BC140991.1 | - | 4 - 5 | 493 | 9055603 | 63 | |
BC145064.1 | - | 5 - 6 | 541 | 9053612 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |