Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nebl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028757.2 | NP_083033.1 | 2 - 3 | 369 | 85 | |
GenBank mRNA | AK004645.1 | - | 2 - 3 | 370 | 85 | |
BC025863.1 | - | 2 - 3 | 344 | 5249766 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |