Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271777.1 | NP_001258706.1 | 1 - 2 | 194 | 61 | |
GenBank mRNA | AK005962.1 | - | 1 - 2 | 198 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |