Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
NM_172537.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
NM_199238.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
NM_199240.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
NM_199241.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014561.mm.1 |
TC0200001866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
NM_172537.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
NM_199238.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
NM_199240.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
NM_199241.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014565.mm.1 |
TC0200001866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
NM_172537.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
NM_199238.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
NM_199240.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
NM_199241.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014563.mm.1 |
TC0200001866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014567.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014567.mm.1 |
NM_199241.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014567.mm.1 |
TC0200001866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
NM_172537.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
NM_199238.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
NM_199240.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
NM_199241.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014562.mm.1 |
TC0200001866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014564.mm.1 |
NM_172537.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014564.mm.1 |
NM_199239.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014564.mm.1 |
TC0200001866.mm.1 |
MTA TC |