Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146073.3 | NP_666185.3 | 1 - 2 | 1091 | 64 | |
GenBank mRNA | AK041496.1 | - | 1 - 2 | 774 | 64 | |
AK046719.1 | - | 1 - 2 | 1079 | 64 | ||
AK080845.1 | - | 1 - 2 | 1095 | 64 | ||
BC059814.1 | - | 1 - 2 | 1134 | 6813525 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |