Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200012044.mm.1 |
NM_021275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012044.mm.1 |
NM_021275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012044.mm.1 |
TC0200001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012042.mm.1 |
NM_021275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012042.mm.1 |
NM_021275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012042.mm.1 |
TC0200001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012039.mm.1 |
NM_021275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012039.mm.1 |
NM_021275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012039.mm.1 |
TC0200001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012040.mm.1 |
NM_021275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012040.mm.1 |
NM_021275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012040.mm.1 |
TC0200001608.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200012043.mm.1 |
NM_021275.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012043.mm.1 |
NM_021275.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200012043.mm.1 |
TC0200001608.mm.1 |
MTA TC |