Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016856.3 | NP_058552.1 | 2 - 3 | 338 | 72 | |
GenBank mRNA | AF012822.1 | - | 2 - 3 | 324 | 72 | |
AK054372.1 | - | 2 - 3 | 321 | 72 | ||
AK140869.1 | - | 2 - 3 | 320 | 72 | ||
AK141325.1 | - | 2 - 3 | 320 | 72 | ||
AK147746.1 | - | 2 - 3 | 339 | 72 | ||
BC013628.1 | - | 2 - 3 | 313 | 3497854 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |