Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029901.2 | NP_084177.2 | 5 - 6 | 646 | 144 | |
GenBank mRNA | AB178898.1 | - | 5 - 6 | 572 | 144 | |
AK020439.1 | - | 5 - 6 | 646 | 144 | ||
AY742217.1 | - | 5 - 6 | 572 | 144 | ||
BC013531.1 | - | 5 - 6 | 622 | 4238756 | 144 | |
BC061057.1 | - | 5 - 6 | 642 | 30305353 | 144 | |
BC091761.1 | - | 5 - 6 | 622 | 5044038 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |