Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146047.2 | NP_666159.2 | 10 - 11 | 1379 | 122 | |
GenBank mRNA | AK032295.1 | - | 10 - 11 | 1343 | 122 | |
AK048357.1 | - | 10 - 11 | 1276 | 122 | ||
AK156203.1 | - | 10 - 11 | 1270 | 122 | ||
AK165939.1 | - | 10 - 11 | 1240 | 122 | ||
AK181076.1 | - | 2 - 3 | 101 | 122 | ||
BC026562.1 | - | 10 - 11 | 1080 | 5028264 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |