Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139877.1 | - | 5 - 6 | 576 | 122 | |
BC049191.1 | - | 14 - 15 | 1995 | 6476607 | 122 | |
BC068151.1 | - | 13 - 14 | 2191 | 30534873 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |