Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172824.3 | NP_766412.2 | 9 - 10 | 1498 | 67 | |
GenBank mRNA | AK029725.1 | - | 8 - 9 | 2630 | 67 | |
DV661299.1 | - | 2 - 3 | 284 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |