Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026254.2 | NP_080530.2 | 4 - 5 | 583 | 79 | |
GenBank mRNA | AK155142.1 | - | 4 - 5 | 604 | 79 | |
AK171591.1 | - | 4 - 5 | 491 | 79 | ||
BC031706.1 | - | 4 - 5 | 583 | 4194266 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |