Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175111.3 | NP_780320.2 | 2 - 3 | 343 | 81 | |
GenBank mRNA | AA879889.1 | - | 2 - 3 | 345 | 81 | |
AK017731.1 | - | 2 - 3 | 309 | 81 | ||
AK028824.1 | - | 2 - 3 | 324 | 81 | ||
AK049404.1 | - | 2 - 3 | 321 | 81 | ||
AK076189.1 | - | 2 - 3 | 310 | 81 | ||
AK089941.1 | - | 2 - 3 | 358 | 81 | ||
AK138906.1 | - | 2 - 3 | 323 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |