Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081172.2 | NP_001074641.1 | 11 - 12 | 1174 | 74 | |
GenBank mRNA | AK122409.1 | - | 8 - 9 | 800 | 74 | |
BC158060.1 | - | 11 - 12 | 1138 | 9007360 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |