Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cast | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301153.1 | NP_001288082.1 | 30 - 31 | 2570 | 80 | |
NM_001301155.1 | NP_001288084.1 | 28 - 29 | 2298 | 80 | ||
NM_001301156.1 | NP_001288085.1 | 27 - 28 | 2244 | 80 | ||
NM_001301157.1 | NP_001288086.1 | 26 - 27 | 2208 | 80 | ||
NM_001301158.1 | NP_001288087.1 | 15 - 16 | 1375 | 80 | ||
NM_001301160.1 | NP_001288089.1 | 15 - 16 | 1333 | 80 | ||
NM_001301181.1 | NP_001288110.1 | 15 - 16 | 1378 | 80 | ||
NM_009817.4 | NP_033947.1 | 29 - 30 | 2330 | 80 | ||
GenBank mRNA | AB026997.1 | - | 30 - 31 | 2569 | 80 | |
AF190151.1 | - | 15 - 16 | 1276 | 80 | ||
AF190152.1 | - | 15 - 16 | 1321 | 80 | ||
AK028843.1 | - | 29 - 30 | 2292 | 80 | ||
AK029293.1 | - | 28 - 29 | 2214 | 80 | ||
AK036576.1 | - | 27 - 28 | 2115 | 80 | ||
AK040688.1 | - | 5 - 6 | 376 | 80 | ||
AK083310.1 | - | 27 - 28 | 2147 | 80 | ||
AK083395.1 | - | 27 - 28 | 2136 | 80 | ||
AK089108.1 | - | 27 - 28 | 2142 | 80 | ||
AK161284.1 | - | 15 - 16 | 1306 | 80 | ||
BC010603.1 | - | 26 - 27 | 2062 | 3710078 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |