Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030711.4 | NP_109636.1 | 15 - 16 | 2322 | 70 | |
GenBank mRNA | AB047552.1 | - | 14 - 15 | 2310 | 70 | |
AF227511.1 | - | 14 - 15 | 2298 | 70 | ||
AK029841.1 | - | 15 - 16 | 2321 | 70 | ||
AK030329.1 | - | 15 - 16 | 2323 | 70 | ||
AK036559.1 | - | 2323 | 70 | |||
AK039505.1 | - | 80 | 70 | |||
AK040350.1 | - | 12 - 13 | 1540 | 70 | ||
AK159406.1 | - | 15 - 16 | 2323 | 70 | ||
BC046610.1 | - | 15 - 16 | 2328 | 6397585 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |