Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177307.3 | NP_796281.1 | 8 - 9 | 1119 | 79 | |
GenBank mRNA | AK028950.1 | - | 8 - 9 | 1120 | 79 | |
AK037192.1 | - | 8 - 9 | 1120 | 79 | ||
BC145755.1 | - | 8 - 9 | 40131058 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |