Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098476.2 | NP_001091946.1 | 2 - 3 | 248 | 101 | |
NM_178700.5 | NP_848815.2 | 2 - 3 | 576 | 101 | ||
GenBank mRNA | AK044889.1 | - | 2 - 3 | 577 | 101 | |
AK045986.1 | - | 2 - 3 | 530 | 101 | ||
AK077782.1 | - | 2 - 3 | 577 | 101 | ||
AK089014.1 | - | 1 - 2 | 112 | 101 | ||
AK168385.1 | - | 2 - 3 | 219 | 101 | ||
AK168459.1 | - | 2 - 3 | 219 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |