Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025568.2 | NP_001020739.1 | 13 - 14 | 1955 | 87 | |
NM_001159952.1 | NP_001153424.1 | 12 - 13 | 1928 | 87 | ||
NM_001159953.1 | NP_001153425.1 | 13 - 14 | 1955 | 87 | ||
NM_001159955.1 | NP_001153427.1 | 14 - 15 | 1517 | 87 | ||
NM_001159956.1 | NP_001153428.1 | 12 - 13 | 1374 | 87 | ||
NM_001159957.1 | NP_001153429.1 | 13 - 14 | 1401 | 87 | ||
NM_001159960.1 | NP_001153432.1 | 14 - 15 | 1829 | 87 | ||
NM_011054.4 | NP_035184.1 | 14 - 15 | 1932 | 87 | ||
GenBank mRNA | AK014887.1 | - | 12 - 13 | 1345 | 87 | |
AK015375.1 | - | 14 - 15 | 1486 | 87 | ||
AK029531.1 | - | 13 - 14 | 1399 | 87 | ||
AK030423.1 | - | 13 - 14 | 1957 | 87 | ||
AK077123.1 | - | 12 - 13 | 1323 | 87 | ||
AK082180.1 | - | 13 - 14 | 1942 | 87 | ||
AK082497.1 | - | 13 - 14 | 1740 | 87 | ||
AK133243.1 | - | 14 - 15 | 1501 | 87 | ||
AK139923.1 | - | 12 - 13 | 1902 | 87 | ||
BC032277.1 | - | 13 - 14 | 1884 | 5400689 | 87 | |
L76944.1 | - | 13 - 14 | 1585 | 87 | ||
L76946.1 | - | 14 - 15 | 1931 | 87 | ||
L76947.1 | - | 13 - 14 | 1418 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |