Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172338.2 | NP_758841.1 | 9 - 10 | 1407 | 67 | |
GenBank mRNA | AK028692.1 | - | 9 - 10 | 1412 | 67 | |
AK051048.1 | - | 9 - 10 | 1408 | 67 | ||
AK122407.1 | - | 1654 | 67 | |||
AK154946.1 | - | 1428 | 67 | |||
AK164023.1 | - | 9 - 10 | 1423 | 67 | ||
BC046424.1 | - | 3 - 4 | 402 | 3989412 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |