Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039200.2 | NP_001034289.1 | 2 - 3 | 257 | 72 | |
NM_146157.4 | NP_666269.2 | 2 - 3 | 257 | 72 | ||
GenBank mRNA | AK049061.1 | - | 2 - 3 | 248 | 72 | |
AK077496.1 | - | 2 - 3 | 247 | 72 | ||
AK089289.1 | - | 2 - 3 | 236 | 72 | ||
AK129052.1 | - | 2 - 3 | 210 | 72 | ||
AK138637.1 | - | 2 - 3 | 229 | 72 | ||
AK161677.1 | - | 2 - 3 | 219 | 72 | ||
AK170739.1 | - | 2 - 3 | 236 | 72 | ||
BC048948.1 | - | 15 - 16 | 72 | |||
BC129804.1 | - | 2 - 3 | 244 | 40087024 | 72 | |
BC139366.1 | - | 2 - 3 | 243 | 8862388 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |