Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010934.4 | NP_035064.1 | 1 - 2 | 104 | 121 | |
GenBank mRNA | AK132397.1 | - | 1 - 2 | 105 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mus musculus Probe spans exons |